Assay Details
| Gene Symbol: sec14l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126793.1 | NP_001120265.1 | 1 - 2 | 83 | 71 | |
| GenBank mRNA | BC160512.1 | - | 1 - 2 | 83 | 71 | |
More Information
Set Membership: |
|

sec14l3
Genomic Map