Assay Details
| Gene Symbol: hpse | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126795.1 | NP_001120267.1 | 1 - 2 | 437 | 75 | |
| XM_012955923.2 | XP_012811377.1 | 2 - 3 | 442 | 75 | ||
| XM_012955930.2 | XP_012811384.1 | 2 - 3 | 442 | 75 | ||
| GenBank mRNA | BC160516.1 | - | 1 - 2 | 437 | 75 | |
More Information
Set Membership: |
|

hpse
Genomic Map