Assay Details
Gene Symbol: pigo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126911.1 | NP_001120383.1 | 8 - 9 | 3186 | 59 | |
GenBank mRNA | BC161087.1 | - | 8 - 9 | 3186 | 59 | |
CT025384.2 | - | 8 - 9 | 3158 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
