Assay Details
Gene Symbol: cnnm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126912.2 | NP_001120384.2 | 1 - 2 | 1571 | 69 | |
XM_012966082.2 | XP_012821536.1 | 1 - 2 | 2203 | 69 | ||
XM_012966083.2 | XP_012821537.1 | 1 - 2 | 2204 | 69 | ||
XM_012966084.2 | XP_012821538.1 | 1 - 2 | 2204 | 69 | ||
GenBank mRNA | BC161088.1 | - | 1 - 2 | 1571 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
