Assay Details
| Gene Symbol: cox19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126977.2 | NP_001120449.1 | 1 - 2 | 165 | 63 | |
| GenBank mRNA | CU075817.1 | - | 1 - 2 | 119 | 63 | |
More Information
Set Membership: |
|

cox19
Genomic Map