Assay Details
Gene Symbol: hs2st1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126991.1 | NP_001120463.1 | 4 - 5 | 767 | 62 | |
XM_018092933.1 | XP_017948422.1 | 4 - 5 | 855 | 62 | ||
GenBank mRNA | BC161283.1 | - | 4 - 5 | 767 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
