Assay Details
| Gene Symbol: cnrip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127084.1 | NP_001120556.1 | 2 - 3 | 392 | 107 | |
| GenBank mRNA | BC161501.1 | - | 2 - 3 | 392 | 107 | |
More Information
Set Membership: |
|

cnrip1
Genomic Map