Assay Details
| Gene Symbol: pyroxd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127086.1 | NP_001120558.1 | 185 | 67 | ||
| GenBank mRNA | BC161508.1 | - | 185 | 67 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pyroxd1| Gene Symbol: pyroxd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127086.1 | NP_001120558.1 | 185 | 67 | ||
| GenBank mRNA | BC161508.1 | - | 185 | 67 | ||
Set Membership: |
|