Assay Details
Gene Symbol: med22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127090.1 | NP_001120562.1 | 293 | 57 | ||
GenBank mRNA | BC161518.1 | - | 293 | 57 | ||
CT025325.2 | - | 3 - 4 | 269 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Molecular Function -



