Assay Details
| Gene Symbol: grk7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127101.1 | NP_001120573.1 | 1 - 2 | 750 | 69 | |
| GenBank mRNA | BC161545.1 | - | 1 - 2 | 750 | 69 | |
More Information
Set Membership: |
|

grk7
Genomic Map