Assay Details
Gene Symbol: mettl24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127109.1 | NP_001120581.1 | 1 - 2 | 375 | 71 | |
GenBank mRNA | BC161568.1 | - | 1 - 2 | 375 | 71 | |
CU075472.1 | - | 1 - 2 | 429 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
