Assay Details
| Gene Symbol: acot9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127117.1 | NP_001120589.1 | 13 - 14 | 1086 | 68 | |
| XM_012956501.2 | XP_012811955.2 | 14 - 15 | 1192 | 68 | ||
| XM_012956502.2 | XP_012811956.2 | 13 - 14 | 1165 | 68 | ||
| XM_012956503.2 | XP_012811957.1 | 14 - 15 | 1294 | 68 | ||
| XM_012956504.2 | XP_012811958.1 | 13 - 14 | 1266 | 68 | ||
| GenBank mRNA | BC161600.1 | - | 13 - 14 | 1086 | 68 | |
More Information
Set Membership: |
|

acot9
Genomic Map