Assay Details
| Gene Symbol: stx10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127158.1 | NP_001120630.1 | 5 - 6 | 774 | 97 | |
| GenBank mRNA | BC161783.1 | - | 5 - 6 | 774 | 97 | |
| CT025172.2 | - | 5 - 6 | 501 | 97 | ||
More Information
Set Membership: |
|

stx10
Genomic Map