Assay Details
| Gene Symbol: znf800 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127423.1 | NP_001120895.1 | 2 - 3 | 167 | 74 | |
| GenBank mRNA | BC166214.1 | - | 2 - 3 | 167 | 74 | |
More Information
Set Membership: |
|

znf800
Genomic Map