Assay Details
| Gene Symbol: pknox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127427.1 | NP_001120899.1 | 7 - 8 | 978 | 59 | |
| GenBank mRNA | BC166172.1 | - | 7 - 8 | 978 | 59 | |
More Information
Set Membership: |
|

pknox2
Genomic Map