Assay Details
| Gene Symbol: hnrnph3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127435.1 | NP_001120907.1 | 9 - 10 | 1107 | 99 | |
| XM_012966113.2 | XP_012821567.1 | 9 - 10 | 1199 | 99 | ||
| XM_012966114.2 | XP_012821568.1 | 9 - 10 | 1200 | 99 | ||
| XM_012966115.2 | XP_012821569.1 | 9 - 10 | 1202 | 99 | ||
| XM_018095280.1 | XP_017950769.1 | 9 - 10 | 1069 | 99 | ||
| GenBank mRNA | BC166302.1 | - | 9 - 10 | 1107 | 99 | |
| BC168807.1 | - | 8 - 9 | 1058 | 99 | ||
More Information
Set Membership: |
|

hnrnph3
Genomic Map