Assay Details
Gene Symbol: picalm.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127945.1 | NP_001121417.1 | 1 - 2 | 433 | 86 | |
GenBank mRNA | BC166117.1 | - | 1 - 2 | 433 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
