Assay Details
Gene Symbol: nomo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127971.1 | NP_001121443.1 | 27 - 28 | 3281 | 103 | |
GenBank mRNA | BC166195.1 | - | 27 - 28 | 3281 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
