Assay Details
| Gene Symbol: nomo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127971.1 | NP_001121443.1 | 7 - 8 | 795 | 103 | |
| GenBank mRNA | BC166195.1 | - | 7 - 8 | 795 | 103 | |
More Information
Set Membership: |
|

nomo3
Genomic Map