Assay Details
| Gene Symbol: tmem59l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128001.1 | NP_001121473.1 | 6 - 7 | 787 | 79 | |
| GenBank mRNA | BC135309.1 | - | 5 - 6 | 695 | 79 | |
| BC166245.1 | - | 6 - 7 | 787 | 79 | ||
| BC167590.1 | - | 6 - 7 | 827 | 79 | ||
More Information
Set Membership: |
|

tmem59l
Genomic Map