Assay Details
Gene Symbol: plpp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128006.2 | NP_001121478.1 | 3 - 4 | 279 | 69 | |
GenBank mRNA | AL888583.2 | - | 3 - 4 | 229 | 69 | |
BC166252.1 | - | 3 - 4 | 232 | 69 | ||
BC168452.1 | - | 2 - 3 | 166 | 69 | ||
CU024953.1 | - | 2 - 3 | 208 | 69 | ||
CX917894.1 | - | 3 - 4 | 270 | 69 | ||
CX928579.1 | - | 3 - 4 | 279 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
