Assay Details
Gene Symbol: plpp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128006.2 | NP_001121478.1 | 6 - 7 | 557 | 97 | |
GenBank mRNA | AL888583.2 | - | 6 - 7 | 507 | 97 | |
BC166252.1 | - | 6 - 7 | 510 | 97 | ||
BC168452.1 | - | 5 - 6 | 444 | 97 | ||
CU024953.1 | - | 5 - 6 | 486 | 97 | ||
CX917894.1 | - | 6 - 7 | 548 | 97 | ||
CX928579.1 | - | 6 - 7 | 557 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
