Assay Details
| Gene Symbol: stard7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128008.1 | NP_001121480.1 | 1 - 2 | 274 | 97 | |
| GenBank mRNA | BC166254.1 | - | 1 - 2 | 274 | 97 | |
More Information
Set Membership: |
|

stard7
Genomic Map