Assay Details
Gene Symbol: fam234a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128045.1 | NP_001121517.1 | 10 - 11 | 1478 | 67 | |
XM_012970131.2 | XP_012825585.1 | 10 - 11 | 1507 | 67 | ||
XM_012970133.2 | XP_012825587.1 | 10 - 11 | 1503 | 67 | ||
XM_018097045.1 | XP_017952534.1 | 10 - 11 | 1413 | 67 | ||
GenBank mRNA | BC166371.1 | - | 10 - 11 | 1478 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
