Assay Details
Gene Symbol: sec62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128046.1 | NP_001121518.1 | 6 - 7 | 613 | 74 | |
XM_018093802.1 | XP_017949291.1 | 6 - 7 | 646 | 74 | ||
GenBank mRNA | BC166373.1 | - | 6 - 7 | 613 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
