Assay Details
| Gene Symbol: pgs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128054.2 | NP_001121526.2 | 7 - 8 | 1330 | 79 | |
| XM_018097672.1 | XP_017953161.1 | 8 - 9 | 1533 | 79 | ||
| GenBank mRNA | BC166392.1 | - | 7 - 8 | 1330 | 79 | |
| CX885342.1 | - | 3 - 4 | 539 | 79 | ||
| EL826393.1 | - | 3 - 4 | 561 | 79 | ||
More Information
Set Membership: |
|

pgs1
Genomic Map