Assay Details
Gene Symbol: abhd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128647.2 | NP_001122119.2 | 3 - 4 | 321 | 61 | |
XM_018091329.1 | XP_017946818.1 | 4 - 5 | 415 | 61 | ||
GenBank mRNA | BC121505.1 | - | 3 - 4 | 321 | 61 | |
BC167663.1 | - | 3 - 4 | 322 | 61 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_018091329.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
