Assay Details
Gene Symbol: zhx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128662.1 | NP_001122134.1 | 2 - 3 | 2565 | 93 | |
XM_012964171.2 | XP_012819625.1 | 3 - 4 | 2965 | 93 | ||
XM_012964172.2 | XP_012819626.1 | 3 - 4 | 2806 | 93 | ||
XM_012964173.2 | XP_012819627.1 | 3 - 4 | 2784 | 93 | ||
XM_018089649.1 | XP_017945138.1 | 2 - 3 | 2892 | 93 | ||
GenBank mRNA | BC167629.1 | - | 2 - 3 | 2565 | 93 | |
CU075533.1 | - | 3 - 4 | 2700 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
