Assay Details
Gene Symbol: rspo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129773.1 | NP_001123245.1 | 3 - 4 | 453 | 74 | |
GenBank mRNA | AY753199.2 | - | 3 - 4 | 453 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
