Assay Details
| Gene Symbol: fzd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129910.1 | NP_001123382.1 | 778 | 57 | ||
| GenBank mRNA | BC124561.1 | - | 735 | 57 | ||
| BC167603.1 | - | 778 | 57 | |||
More Information
Set Membership: |
|

fzd3
Genomic Map