Assay Details
Gene Symbol: pou5f3.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129934.1 | NP_001123406.1 | 1 - 2 | 976 | 96 | |
GenBank mRNA | BC167674.1 | - | 1 - 2 | 976 | 96 | |
BC167702.1 | - | 1 - 2 | 963 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
