Assay Details
Gene Symbol: pou5f3.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129934.1 | NP_001123406.1 | 2 | 1084 | 102 | |
GenBank mRNA | BC167674.1 | - | 2 | 1084 | 102 | |
BC167702.1 | - | 2 | 1071 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
