Assay Details
| Gene Symbol: twf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129945.1 | NP_001123417.1 | 7 - 8 | 862 | 91 | |
| GenBank mRNA | BC118802.1 | - | 7 - 8 | 862 | 91 | |
More Information
Set Membership: |
|

twf2
Genomic Map