Assay Details
| Gene Symbol: elp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130214.1 | NP_001123686.1 | 11 - 12 | 1492 | 67 | |
| XM_012956928.1 | XP_012812382.1 | 11 - 12 | 1278 | 67 | ||
| XM_012956932.1 | XP_012812386.1 | 11 - 12 | 1278 | 67 | ||
| XM_012956940.1 | XP_012812394.1 | 10 - 11 | 1066 | 67 | ||
| GenBank mRNA | BC121944.1 | - | 10 - 11 | 1195 | 67 | |
| BC166239.1 | - | 11 - 12 | 1492 | 67 | ||
More Information
Set Membership: |
|

elp1
Genomic Map