Assay Details
Gene Symbol: pqlc2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130218.1 | NP_001123690.1 | 4 - 5 | 635 | 107 | |
GenBank mRNA | BC167268.1 | - | 4 - 5 | 635 | 107 | |
BC167667.1 | - | 4 - 5 | 635 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
