Assay Details
Gene Symbol: pqlc2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130218.1 | NP_001123690.1 | 5 - 6 | 726 | 64 | |
GenBank mRNA | BC167268.1 | - | 5 - 6 | 726 | 64 | |
BC167667.1 | - | 5 - 6 | 726 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
