Assay Details
| Gene Symbol: cry4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130234.1 | NP_001123706.1 | 3 - 4 | 591 | 74 | |
| GenBank mRNA | AB534559.1 | - | 2 - 3 | 538 | 74 | |
| BC167313.1 | - | 3 - 4 | 591 | 74 | ||
More Information
Set Membership: |
|

cry4
Genomic Map