Assay Details
Gene Symbol: nme6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130237.1 | NP_001123709.1 | 4 - 5 | 417 | 65 | |
XM_012958647.2 | XP_012814101.1 | 5 - 6 | 606 | 65 | ||
XM_018091950.1 | XP_017947439.1 | 5 - 6 | 429 | 65 | ||
GenBank mRNA | BC167315.1 | - | 4 - 5 | 417 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
