Assay Details
Gene Symbol: spart | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130255.1 | NP_001123727.1 | 1 - 2 | 829 | 87 | |
XM_012956537.2 | XP_012811991.1 | 2 - 3 | 997 | 87 | ||
GenBank mRNA | BC167368.1 | - | 1 - 2 | 829 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
