Assay Details
Gene Symbol: spart | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130255.1 | NP_001123727.1 | 3 - 4 | 1152 | 63 | |
XM_012956537.2 | XP_012811991.1 | 4 - 5 | 1320 | 63 | ||
GenBank mRNA | BC167368.1 | - | 3 - 4 | 1152 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
