Assay Details
| Gene Symbol: bcr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130320.1 | NP_001123792.1 | 1 - 2 | 1606 | 60 | |
| GenBank mRNA | BC167580.1 | - | 1 - 2 | 1606 | 60 | |
More Information
Set Membership: |
|

bcr
Genomic Map