Assay Details
Gene Symbol: snrk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130352.1 | NP_001123824.1 | 4 - 5 | 1340 | 69 | |
GenBank mRNA | BC167632.1 | - | 4 - 5 | 1340 | 69 | |
CT030415.1 | - | 4 - 5 | 1225 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
