Assay Details
Gene Symbol: pou5f3.3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130364.1 | NP_001123836.1 | 906 | 64 | ||
GenBank mRNA | BC167672.1 | - | 906 | 64 | ||
CU075527.1 | - | 884 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
