Assay Details
| Gene Symbol: rgs16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130383.1 | NP_001123855.1 | 4 - 5 | 379 | 70 | |
| GenBank mRNA | BC167734.1 | - | 4 - 5 | 379 | 70 | |
More Information
Set Membership: |
|

rgs16
Genomic Map