Assay Details
Gene Symbol: gdf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135117.1 | NP_001128589.1 | 1 - 2 | 620 | 86 | |
GenBank mRNA | AB236907.1 | - | 1 - 2 | 620 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
