Assay Details
Gene Symbol: aifm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142019.1 | NP_001135491.1 | 4 - 5 | 519 | 71 | |
XM_018095294.1 | XP_017950783.1 | 3 - 4 | 529 | 71 | ||
GenBank mRNA | BC167890.1 | - | 4 - 5 | 519 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
