Assay Details
| Gene Symbol: lclat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142045.1 | NP_001135517.1 | 4 - 5 | 643 | 75 | |
| XM_012962792.1 | XP_012818246.1 | 4 - 5 | 642 | 75 | ||
| GenBank mRNA | BC167939.1 | - | 4 - 5 | 643 | 75 | |
More Information
Set Membership: |
|

lclat1
Genomic Map