Assay Details
| Gene Symbol: prkaa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142082.1 | NP_001135554.1 | 1 - 2 | 102 | 63 | |
| GenBank mRNA | BC168035.1 | - | 1 - 2 | 102 | 63 | |
More Information
Set Membership: |
|

prkaa2
Genomic Map