Assay Details
Gene Symbol: atp5if1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142093.1 | NP_001135565.1 | 1 - 2 | 99 | 82 | |
GenBank mRNA | BC168056.1 | - | 1 - 2 | 99 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
