Assay Details
| Gene Symbol: atp5if1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142093.1 | NP_001135565.1 | 2 - 3 | 211 | 75 | |
| GenBank mRNA | BC168056.1 | - | 2 - 3 | 211 | 75 | |
More Information
Set Membership: |
|

atp5if1
Genomic Map